Tlačiť
Test tubes with screw caps
Dodávateľ: BOETTGER PAUL
01.073.2000 01.083.1400 01.083.2400 01.063.1000 01.063.2000 01.073.1000 01.073.3100 01.083.3400 01.093.1000 01.063.3100
212-0512EA 296.43 EUR
212-0512 212-0514 212-0515 212-0508 212-0509 212-0511 212-0513 212-0516 212-0517 212-0510
Test tubes with screw caps
Zkumavky Zkumavky na vzorky
PP, transparent.
  • Good chemical and temperature resistance
  • Tight sealing screw cap made from PE
  • Available with enclosed or fitted screw cap, standard-hygienic or sterile
For safe transportation and storage of samples.