Tlačiť

Hydrogenfosforečnan diamonný

Dodávateľ: Lach-ner S.R.O.

Synonymá: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

30059CP0G1000 30059CP0G0500
LACH30059CP0G1000EA 0 EUR
LACH30059CP0G1000 LACH30059CP0G0500
Hydrogenfosforečnan diamonný
Hydrogenfosforečnan diamonný
Vzorec: (NH₄)₂HPO₄
Mw: 132.06
Teplota tavenia: 185
Hustota: 1.619 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00010891
Číslo CAS: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR