Tlačiť

Hydrogenfosforečnan didraselný

Dodávateľ: Lach-ner S.R.O.

Synonymá: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

30060AP0G0500 30060AP0G02501 30060AP0G05001 30060AP0G1000
LACH30060AP0G0500EA 20.34 EUR
LACH30060AP0G0500 LACH30060AP0G02501 LACH30060AP0G05001 LACH30060AP0G1000
Hydrogenfosforečnan didraselný
Hydrogenfosforečnan didraselný
Vzorec: K₂HPO₄
Teplota tavenia: 340 °C
Hustota: 2,3 g/cm³ (20 °C)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00011383
Číslo CAS: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR