Tlačiť
ESD safety shoes, lace-up, Compositelite™ ESD FC02
Dodávateľ: Portwest
FC02BKR37 FC02BKR39 FC02BKR40 FC02BKR42 FC02BKR47
114-4353EA 80.22 EUR
114-4353 114-4355 114-4356 114-4358 114-4363
ESD safety shoes, lace-up, Compositelite™ ESD FC02
Obuv
Metal-free, anti-static ESD safety shoes featuring a split leather upper plus a dual-density PU sole, resistant to oil.
  • Padded collar and tongue
  • Shock-absorbent heel
  • Composite toecap (200 J)
Suitable for ESD environments.
Colour: Black
Certifikácie: EN 61340 (ESD); EN 20345:2011 + A1:2007; S1, SRC; IEC 61340-4-3.