Tlačiť
Tracking system for single and multi channel pipettes, Trackman™
Dodávateľ: GILSON
F70301 F70261 F70105
GILSF70301EA 1016.45 EUR
GILSF70301 GILSF70261 GILSF70105
Tracking system for single and multi channel pipettes, Trackman™
Pipety Micropipettes
This system with 16 programming possibilities for pipetting patterns (rows, columns, and more.) is designed specifically to keep users on track as they pipette from one microplate to another. It simplifies work, improves productivity and helps avoid pipetting errors and cross-contamination.
  • Light-weight and ergonomic
  • Intuitive software that requires no training
  • Avoid pipetting errors and cross-contamination
  • Timer function enables hands-free operation
  • Optional foot pedal available for hands free control
  • Long lasting battery allows cordless operation for up to eight hours
Can be used with both 96 and 384 well microplates, and single and multichannel pipettes. Can also be used with 0,5, 1,5 and 2,0 ml tubes with the optional tube holder accessory.
Informace o dodávce: System includes power supply with AC adapter, cable, three reagent reservoirs.