Tris[N,N-bis(trimethylsilyl)amide]neodymium(III) ≥98%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
44395.06
AB204475.0001
44395.06EA
607.65
EUR
44395.06
ABCRAB204475.0001
Tris[N,N-bis(trimethylsilyl)amide]neodymium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amide]neodymium(III)
Purity: 98%
CAS No.: 148274-47-9
Molecular Formula: C18H54N3NdSi6
Formula Weight: 625.39
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Pale purple
Melting Point: 156-159°
UN No.: 3396
MDL No.: MFCD03427089
Vzorec:
C₁₈H₅₄N₃NdSi₆ Mw: 625.39 g/mol Teplota tavenia: 156…159 °C Skladovacia teplota: Ambient |
Číslo MDL:
MFCD03427089 Číslo CAS: 148274-47-9 OSN: 3396 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...