Tlačiť
Culture shake flasks, with three baffles
Dodávateľ: BELLCO GLASS
2540-00125 2540-00250 2540-00300 2540-00500 2540-01000 2540-02000
BELC2540-00125EA 1011.42 EUR
BELC2540-00125 BELC2540-00250 BELC2540-00300 BELC2540-00500 BELC2540-01000 BELC2540-02000
Culture shake flasks, with three baffles
Baňky Baňky kultivační
Flasks, made of type 1 class A borosilicate glass, with DeLong® style neck, these shake flasks are the basic tool for preliminary fermentation and bacterial growth studies.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes