Tlačiť
Separating funnels, Squibb
Dodávateľ: LENZ LABORGLAS
04018049 04018058 04018149 04018137 04018037
LENZ04018049EA 237.46 EUR
LENZ04018049 LENZ04018058 LENZ04018149 LENZ04018137 LENZ04018037
Separating funnels, Squibb
Nálevky Nálevky dělící
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids