Tlačiť
Lens cleaning tissues, grade 913
Dodávateľ: Ahlstrom Munksjö
0913F00022 0913F00023
FIOR0913F00022EA 3.22 EUR
FIOR0913F00022 FIOR0913F00023
Lens cleaning tissues, grade 913
Ubousky/Utěrky Utěrky jednorázové a ručníky Papíry na čištění čoček
These tissues are suitable for cleaning various optical lenses such as objectives, microscope, binoculars, magnifying glasses, spectacles as well as optical fibre filaments.
  • Made from 100% manila
Thickness: 0,04 mm
Weight: 12 g/m²