Tlačiť
Qualitative filter papers
Dodávateľ: Ahlstrom Munksjö
0111A00002 0111A00005 0111A00006 0113A00005 0113A00006 0113A00012 0113P00024 0113P00029
FIOR0111A00002EA 3.94 EUR
FIOR0111A00002 FIOR0111A00005 FIOR0111A00006 FIOR0113A00005 FIOR0113A00006 FIOR0113A00012 FIOR0113P00024 FIOR0113P00029
Qualitative filter papers
Filtry Papíry filtrační Papír kvalitativní filtrační
These filter papers are used in chemical, food and beverage industries, also for atmospheric analysis, essential and industrial oil applications.
  • They are made of cellulose with an alpha content
  • Extremely pure with an ash content of 0,06%
Grade 111: Fast filtration used for coarse and voluminous precipitates like hydroxides and sulphides.
Grade 113: Medium filtration suitable for fast filtration of liquids with fine particles.