Tlačiť
Columns, MicroBore
Dodávateľ: VARIAN
PL1312-1300 PL1312-1301 PL1312-1500 PL1312-1501 PL1312-1502 PL1312-1503 PL1312-1802 PL1312-1803 PL1312-3300 PL1312-3301 PL1345-1502 PL1345-1503 PL1351-1502 PL1351-1503
VARIPL1312-1300EA 709.99 EUR
VARIPL1312-1300 VARIPL1312-1301 VARIPL1312-1500 VARIPL1312-1501 VARIPL1312-1502 VARIPL1312-1503 VARIPL1312-1802 VARIPL1312-1803 VARIPL1312-3300 VARIPL1312-3301 VARIPL1345-1502 VARIPL1345-1503 VARIPL1351-1502 VARIPL1351-1503
Columns, MicroBore
Kolony chromatografické
The microbore columns are ideal for use with detectors requiring low flow rates such as some mass
spectrometers and with capillary LC systems. Easily accommodates high temperatures for better selectivity and resolution between impurities and the target oligo.
  • Thermally and chemically stable
  • Exceptional resolution and selectivity
  • Contain durable and resilient polymer particles that deliver reproducible results over longer lifetimes
  • High purity levels
  • Gives a highly reproducible material that is free from silanols and heavy metal ions.
This consists of a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic, therefore no bonded phase, alkyl ligand is required for reversed-phase separations. Columns within the extensive product range are suitable for nano/capillary separations, including both bottom-up and top-down proteomics, analytical separations, and preparative purifications.