Tlačiť
Capillary columns, CP-Chirasil-Dex CB
Dodávateľ: VARIAN
CP7502 CP7503 CP7502I5
VARICP7502EA 1493.38 EUR
VARICP7502 VARICP7503 VARICP7502I5
Capillary columns, CP-Chirasil-Dex CB
Kolony chromatografické
The CP-Chirasil-Dex CB phase consists of cyclodextrin directly bonded to dimethylpolysiloxane. This bond prevents the cyclodextrin from migrating to different locations in the surface film, delivering homogeneous enantioselectivity throughout the phase.
  • High resolution across a broad application range
  • Chemically-bonded phase for excellent longevity
  • No need for derivatisation improves productivity
  • Guarantees stability of enantioselectivity
CP-Chirasil-Dex CB permits low elution temperatures of polar compounds and is suitable for all injection techniques.