Tlačiť
Capillary columns, CP-SimDist
Dodávateľ: VARIAN
CP7541 CP7521 CP7522 CP7522I5
VARICP7541EA 1404.73 EUR
VARICP7541 VARICP7521 VARICP7522 VARICP7522I5
Capillary columns, CP-SimDist
Kolony chromatografické
CP-SimDist Fused Silica columns are guaranteed for simulated distillation up to C100.
  • Conforms to ASTM Method D 2887 to provide full compliancy
  • High temperature stationary phase for extended column lifetime
  • Low bleed makes quantitation easier