Tlačiť
SPE columns, CHROMABOND® PCA
Dodávateľ: MACHEREY-NAGEL
730482 730483 730484
MANA730482EA 276.37 EUR
MANA730482 MANA730483 MANA730484
SPE columns, CHROMABOND® PCA
Kolony chromatografické
CHROMABOND® PCA is a propylcarboxylic acid modified silica cation exchanger for SPE.
  • Base material: Silica
  • Weakly acidic cation exchanger (WCX)
Recommended for strong cations like cationic peptides (viscotoxines).