Tlačiť

Ruthenium(III) chloride hydrate

Dodávateľ: abcr
Nebezpečí

Synonymá: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

AB101280.0005 AB101280.0025
ABCRAB101280.0005EA 0 EUR
ABCRAB101280.0005 ABCRAB101280.0025
Ruthenium(III) chloride hydrate
Ruthenium(III) chloride hydrate
Vzorec: Cl₃Ru·xH₂O
Mw: 225.44
Teplota tavenia: > 300 °C
Hustota: 3.11 g/cm³ (20 °C)
Číslo MDL: MFCD00149844
Číslo CAS: 14898-67-0
EINECS: 233-167-5
OSN: 3260

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR