Tlačiť
Transport jugs
Dodávateľ: BOCHEM
8380 8384 8381
216-0011EA 1138.39 EUR
216-0011 216-0013 216-0012
Transport jugs
Láhve Demižóny
Transport jug made of stainless steel 18/10, with spout, handle and lid.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean