Tlačiť
Cleanroom anti-fatigue mats, Ergomat® AB Classic
Dodávateľ: ERGOMAT
ST0404 ST0420 ST0412 ST0406 ST0320 ST0306 ST0304 ST0220 ST0209 ST0206 ST0203 ST0212 ST0303 ST0312 ST0309 ST0409
115-0540EA 456.08 EUR
115-0540 115-0794 115-0793 115-0792 115-0791 115-0790 115-0789 115-0788 115-0787 115-0786 115-0785 115-2946 115-2947 115-2949 115-2948 115-2951
Cleanroom anti-fatigue mats, Ergomat® AB Classic
Rohože Rohože proti únavě
The anti-static Ergomat® AB Classic is made of pure polyurethane and is designed to increase comfort while reducing trip hazards and body pain. It is ideal for use in dry environments or where control of static electricity is needed.
  • Upper side with bubbled surface
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantee durability and ergonomic benefits
  • Solid mat throughout, no hollow bubbles that collapse
  • Anti-static, free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Anthracite
Certifikácie: Meets EN ISO standards