Tlačiť
Hoffmann clips
Dodávateľ: MURRAY S
L141/01 L141/02 L141/03 L141/04 L141/05
MURRL141/01EA 19.57 EUR
MURRL141/01 MURRL141/02 MURRL141/03 MURRL141/04 MURRL141/05
Hoffmann clips
Svorky Svorky trubkové
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
  • Spanning width = Width of gate