Tlačiť
Capillary columns, VF-35ms
Dodávateľ: VARIAN
CP8882 CP8883 CP8884 CP8879 CP8874 CP8875 CP8877 CP8878 CP8873 CP5887 CP5889
VARICP8882EA 1155.64 EUR
VARICP8882 VARICP8883 VARICP8884 VARICP8879 VARICP8874 VARICP8875 VARICP8877 VARICP8878 VARICP8873 VARICP5887 VARICP5889
Capillary columns, VF-35ms
Kolony chromatografické
VF-35ms is a medium polarity column, which is the ideal choice for trace environmental and chemical analyses, and as a confirmation column.
  • Ideal for dual column confirmational analysis for ultimate confidence
  • High maximum temperature for broad applicability
  • Stabilized arylene-modified equivalent of a 35% phenylmethyl phase for longevity
VF-35ms uses FactorFour technology to produce a low bleed, highly stable column with a programmable maximum temperature of 360 °C.