Tlačiť
Nylon manifold; 8 outlets; 1/4" inlet NPT(F)
Dodávateľ: Avantor Fluid Handling
VWR Item#MFLX31522-49
Your Organization's Price:
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
  • Constructed of durable nylon
Constructed of nylon for economic general applications while offering good chemial compatibility.
More Product Information
Other Products In this Family
See all Items in this Product Family