Tlačiť
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 3/32" ID x 10-32 UNF(M); 10/PK
Dodávateľ: Avantor Fluid Handling
VWR Item#MFLX40629-00
Your Organization's Price:
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/32" ID x 10-32 UNF(M)
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
More Product Information
Other Products In this Family
See all Items in this Product Family