Tlačiť
Trypsinising flasks
Dodávateľ: witeg Labortechnik
5.507.305 5.507.308 5.507.306 5.507.303 5.507.304 5.507.307
WITG5.507.305EA 72.1 EUR
WITG5.507.305 WITG5.507.308 WITG5.507.306 WITG5.507.303 WITG5.507.304 WITG5.507.307
Trypsinising flasks
Baňky Baňky trypsinační
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable