Tlačiť
Pycnometer for ice creme tests
Dodávateľ: witeg Labortechnik
3.935.003
WITG3.935.003EA 64.09 EUR
WITG3.935.003
Pycnometer for ice creme tests
Pyknometry
Pycnometer, made of borosilicate glass 3.3.
  • Cap. 20 %
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance