Tlačiť
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
Dodávateľ: SP Industries
WG-3000-7-50
WILMWG-3000-7-50EA 205.09 EUR
WILMWG-3000-7-50
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
Zkumavky Zkumavky pro NMR
High-throughput NMR tubes are made of borosilicate glass Type I, class A.
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable