Tlačiť
WHEATON® COMPLETEPACK MODPACK Vials with Stoppers and Seals, Amber Glass
Dodávateľ: DWK LIFE SCIENCES
W991000A-CPMP W991001A-CPMP W991002A-CPMP W991003A-CPMP W991004A-CPMP W991005A-CPMP W991006A-CPMP W991007A-CPMP W991008A-CPMP W991009A-CPMP W991010A-CPMP W991011A-CPMP W991012A-CPMP W991013A-CPMP W991014A-CPMP W991015A-CPMP W991016A-CPMP W991017A-CPMP
WHEAW991000A-CPMPEA 16827.16 EUR
WHEAW991000A-CPMP WHEAW991001A-CPMP WHEAW991002A-CPMP WHEAW991003A-CPMP WHEAW991004A-CPMP WHEAW991005A-CPMP WHEAW991006A-CPMP WHEAW991007A-CPMP WHEAW991008A-CPMP WHEAW991009A-CPMP WHEAW991010A-CPMP WHEAW991011A-CPMP WHEAW991012A-CPMP WHEAW991013A-CPMP WHEAW991014A-CPMP WHEAW991015A-CPMP WHEAW991016A-CPMP WHEAW991017A-CPMP
WHEATON® COMPLETEPACK MODPACK Vials with Stoppers and Seals, Amber Glass
Vialky
Six inventory matched kits of amber crimp top vials, rubber stoppers and seals that are sterilised and ready-to-use packed into a shipper case.
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Amber glass vials especially for photosensitive contents
Certifikácie: USP <788>/EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers)More Product Information