Tlačiť
Weighing dishes, aluminium, WHEATON®
Dodávateľ: DWK LIFE SCIENCES
370792
WHEA370792EA 185.06 EUR
WHEA370792
Weighing dishes, aluminium, WHEATON®
Lodičky Lodičky navažovací
Ideal for milligram weighing of liquids or solids.
  • Lightweight, easy to clean
  • Good chemical resistance
  • Good temperature resistance