Tlačiť
Filtry odvzdušňovací, SteriVENT™, Whatman™
Dodávateľ: Whatman products (Cytiva)
2103 2107 2108
WHAT2103EA 127.38 EUR
WHAT2103 WHAT2107 WHAT2108
Filtry odvzdušňovací, SteriVENT™, Whatman™
Filtry Filtry odvzdušňovací
SteriVENT™ venting filters with an integral hydrophobic PTFE membrane and PP housing are well suited for sterile venting of vessels and tanks during draining and filling. These filters are capable of allowing clean air to pass into the vessel while preventing microbiological contamination.
  • Industrial air filters for sterile venting
  • Can be autoclaved 50 times without loss of performance; compatible with ethylene oxide (EtO)
  • Integrity can be tested with the water break through (WBT) test
  • Bidirectional flow direction
  • Heat fused sealing
  • Vent on inlet
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable
Ø 36 mm 150 mm 150 mm
Typ krytu Kapsle SteriVENT™ 36 Kapsle SteriVENT™ 150 Kapsle SteriVENT™ 150
Maximální tlak 2 bar 2 bar 2 bar
Vstup/výstup Olivka 3/8" až 1/2" Vroubkovaný nástavec 3/8" až 1/2" 1 1/2" sanitární/1 1/2" sanitární