Tlačiť
Detergent, neodisher LaboClean FT
Dodávateľ: DR WEIGERT
410030_U
WEIG410030_UEA 161.83 EUR
WEIG410030_U
Detergent, neodisher LaboClean FT
Prostředky čistící Detergenty a povrchové čističe
neodisher® LaboClean FT
  • Alkaline liquid concentrate especially suitable for removal of residues of culture media, blood, proteins and other organic materials
  • Components: Potassium hydroxide and sodium hypochlorite
  • Dosage: 3 to 4 ml/L
Pozor: Most of the materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.