Tlačiť
VWR® Square Polycarbonate Media Bottles
Dodávateľ: VWR Collection
VWRI210-000090EA 340.48 EUR
VWRI210-000090 VWRI210-000094 VWRI210-000102 VWRI210-000095 VWRI210-000101 VWRI210-000103 VWRI210-000100 VWRI210-000093 VWRI210-000099 VWRI210-000096 VWRI210-000098 VWRI210-000087 VWRI210-000097 VWRI210-000104 VWRI210-000089 VWRI210-000086 VWRI210-000091 VWRI210-000088 VWRI210-000092 VWRI210-000085
VWR® Square Polycarbonate Media Bottles
Láhve Láhve plastové
VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
  • Made from durable 100% virgin polycarbonate material
  • Suitable for frozen storage down to −130 °C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and closures (HDPE/PP) provided for applicable products
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-
More Product Information