Tlačiť
Dropping bottles, with stopper
Dodávateľ: KAVALIER
632426005050 632426005100 632426005250
VTRB632426005050EA 23.67 EUR
VTRB632426005050 VTRB632426005100 VTRB632426005250
Dropping bottles, with stopper
Láhve Lahvičky kapací
Borosilicate glass 3.3 SIMAX®.
  • Very good temperature resistance
  • Very good chemical resistance