Tlačiť

Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F104]

Dodávateľ: UNITED BIOCHEMICALS
M3887-43D
USBIM3887-43DEA 868.02 EUR
USBIM3887-43D
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F104]
Protilátky
Recommended dilution: Flow Cytometry: 1:10;

Type: Primary
Antigen: MHC Class 2, DQ, DR, Polymorphic
Clonality: monoclonal
Clone: 10F104
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Goat, Human, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR