Tlačiť

Anti-GABA Receptor A, delta Mouse monoclonal antibody [clone: 10F704]

Dodávateľ: UNITED BIOCHEMICALS
G1015-19N4
USBIG1015-19N4EA 669.74 EUR
USBIG1015-19N4
Anti-GABA Receptor A, delta Mouse monoclonal antibody [clone: 10F704]
Protilátky
Recommended dilution: Western Blot:1-10 ?g/ml; Immunochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: GABA Receptor A, delta
Clonality: monoclonal
Clone: 10F704
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR