Tlačiť

Anti-Cleavage Stimulation Factor 2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodávateľ: UNITED BIOCHEMICALS
C7946-70C-APC
USBIC7946-70C-APCEA 1008.21 EUR
USBIC7946-70C-APC
Anti-Cleavage Stimulation Factor 2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Cleavage Stimulation Factor 2
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR