Tlačiť

Anti-Major Histocompatibility Complex Class II DR Alpha Mouse Monoclonal Antibody [clone: 13j65]

Dodávateľ: UNITED BIOCHEMICALS
141369
USBI141369EA 674.14 EUR
USBI141369
Anti-Major Histocompatibility Complex Class II DR Alpha Mouse Monoclonal Antibody [clone: 13j65]
Protilátky
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Major Histocompatibility Complex Class II DR Alpha
Clonality: monoclonal
Clone: 13j65
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR