Tlačiť

Anti-SMS Mouse Monoclonal Antibody [clone: 1G6]

Dodávateľ: UNITED BIOCHEMICALS
133585
USBI133585EA 747.05 EUR
USBI133585
Anti-SMS Mouse Monoclonal Antibody [clone: 1G6]
Protilátky
Type: Primary
Antigen: SMS (spermine synthase)
Clonality: monoclonal
Clone: 1G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR