Tlačiť

Anti-NDN Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: UNITED BIOCHEMICALS
038844-BIOTIN
USBI038844-BIOTINEA 1008.21 EUR
USBI038844-BIOTIN
Anti-NDN Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NDN (necdin, melanoma antigen (MAGE) family member)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR