Tlačiť
Face shield, medical, 710
Dodávateľ: UNIVET
710.00.00.00
UNVT710.00.00.00EA 38.41 EUR
UNVT710.00.00.00
Face shield, medical, 710
Štíty obličejové
This face shield features an adjustable elastic band and an anallergic, interchangeable foam at the front.
  • Interchangeable
  • 90° flip up
Weight: 97 g
EN 166 (3 S CE)