Tlačiť
Molecular weight calibration standards for SEC
Dodávateľ: TOSOH Bioscience
0005211 0006476 0006477 0006216 0006212 0005214 0005202 0005203 0005204 0005205 0005206 0005207 0005208 0005209 0006215 0005213 0005215 0005216 0005217 0005218 0005219 0005220 0005221 0005773 0005210
TOYO0005211EA 503.5 EUR
TOYO0005211 TOYO0006476 TOYO0006477 TOYO0006216 TOYO0006212 TOYO0005214 TOYO0005202 TOYO0005203 TOYO0005204 TOYO0005205 TOYO0005206 TOYO0005207 TOYO0005208 TOYO0005209 TOYO0006215 TOYO0005213 TOYO0005215 TOYO0005216 TOYO0005217 TOYO0005218 TOYO0005219 TOYO0005220 TOYO0005221 TOYO0005773 TOYO0005210
Molecular weight calibration standards for SEC
Standardy Standardy pro chromatografii
These standards consists of bulk quantities of polystyrene and polyethylene oxide standards of varying molar mass and pre-mixed quantities of polystyrene polymers for calibration of GPC columns.