Tlačiť
TELSTAR BiOptima EXA Biological Safety Cabinets, Class II
Dodávateľ: Azbil Telstar Technologies
1738689 1745204
TELR1738689EA 17146 EUR
TELR1738689 TELR1745204
TELSTAR BiOptima EXA Biological Safety Cabinets, Class II
Digestoře Biologické boxy
The ergonomically designed BiOptima Class II microbiological safety cabinets ensure optimum efficiency and high-level energy savings. BiOptima EXA provide operator protection by inflow through the front grill, product protection by HEPA-filtered laminar downflow in the working area (100% exhaust) and environmental protection via HEPA-filtered exhaust air (not included with the cabinet). BiOptima EXA is designed for hazardous airborne application such as volatiles, mists or vapours beside high microbiological and hight toxic applications. Designed according to NSF 49. This model is availableMore Product Information