Tlačiť
Stand-up writing desk/tables, 304 stainless steel
Dodávateľ: TEKNOMEK
DEWR-1SHMS DEWR-2SHMS DEWR-2SHS DEWR-CBMS DEWR-CBS DEWR-MS DEWR-S DEWR-WM
TEKMDEWR-1SHMSEA 0 EUR
TEKMDEWR-1SHMS TEKMDEWR-2SHMS TEKMDEWR-2SHS TEKMDEWR-CBMS TEKMDEWR-CBS TEKMDEWR-MS TEKMDEWR-S TEKMDEWR-WM
Stand-up writing desk/tables, 304 stainless steel
Nábytek kancelářský Stoly
304-grade stainless steel stand up desks with easy access document storage. Manufactured for optimal durability and strength. They are fully welded, with reduce dirt traps to reduce risk. Choose from free-standing, wall mounted or mobile, with a wide choice of shelf options too.
  • 304-grade, EU sourced stainless steel
  • Supplied fully welded, no construction or assembly required
  • Ideal for standing
  • Hygienic design throughout, minimised dirt traps
  • Brushed dull polish finish
  • Choice of mobile or static or wall mounted
  • Choice of no shelf, one shelf or two shelves
More Product Information