Tlačiť
Cole-Parmer® GT-200 Gelation Timers
Dodávateľ: TSC BIOSCIENCES
98930-41 98930-43 98930-49 98930-47 98930-48 98930-46 98930-44
TECN98930-41EA 3743.24 EUR
TECN98930-41 TECN98930-43 TECN98930-49 TECN98930-47 TECN98930-48 TECN98930-46 TECN98930-44
Cole-Parmer® GT-200 Gelation Timers
Časovace/Stopky Časovače digitální
Measure the gelation time of adhesives and resins for quality control.
  • Automatically stops when gelation occurs
  • Audible and visual alarms alert user at the gelation point
  • Simple-to-use push buttons with large LED display
  • Stainless steel plungers remove for easy cleaning
The gelation time is the transition from liquid to solid during polymerisation. Test requires approximately 100 ml of sample in a container that is >1,5" (38 mm) in diameter. Once the measurement is started, the plunger moves up and down. After gelation, the plunger is halted,More Product Information