Tlačiť
Cole-Parmer® Stuart WS-100 Water Stills
Dodávateľ: STUART SCIENTIFIC
99293-00 99295-06
STUA99293-00EA 2666.53 EUR
STUA99293-00 STUA99295-06
Cole-Parmer® Stuart WS-100 Water Stills
Systémy pro výrobu čisté vody Přístroje destilační
Save money while maintaining high performance.
  • Two independent thermostats prevent overheating
  • Features a sturdy metal stand for either wall or bench mounting
  • Operate directly from tap water
This basic economical water still is perfect for general laboratory and educational distillations. Borosilicate glass boiler and condenser provide the highest purity distillate. The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate coldMore Product Information