Tlačiť

Magnesium aluminum silicate NF Type IA

Dodávateľ: Spectrum Chemical

M1021-2.5KG M1021-500GM M1021-12KG M1021-45KGBL
SPCMM1021-2.5KGEA 816.34 EUR
SPCMM1021-2.5KG SPCMM1021-500GM SPCMM1021-12KG SPCMM1021-45KGBL
Magnesium aluminum silicate NF Type IA
Magnesium aluminum silicate

Magnesium Aluminum Silicate, Type IA, NF is used in the pharmaceutical formulation process as an absorbent, anticaking agent, opacifying agent, slip modifier, and an aqueous viscosity increasing agent. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Skladovacia teplota: Ambient
Číslo CAS: 1327-43-1

Order Now

Výsledky špecifikačného testu

Viscosity @ 33 +/- 3 °C 225 - 600 cP
pH of a 5 in 100 Suspension 9.0 - 10.0
Ratio of Content Aluminum / Magnesium 0.5 - 1.2
Acid Demand max. 4.0
Loss On Drying max. 8.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR