Tlačiť

Glycogen, powder

Dodávateľ: Spectrum Chemical

GL160-5GM
SPCMGL160-5GMEA 499.5 EUR
SPCMGL160-5GM
Glycogen, powder
Glycogen

Glycogen, Powder is a multibranched polysaccharide of glucose which represent the primary storage of glucose in the body, as well as serving as an energy storage in fungi and animals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Vzorec: C₂₄H₄₂O₂₁
Mw: 666.579 g/mol
Skladovacia teplota: Refrigerator
Číslo MDL: MFCD00081547
Číslo CAS: 9005-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR