Tlačiť

EDTA dvojsodná sůl dihydrát 99,0 FCC

Dodávateľ: Spectrum Chemical

Synonymá: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

E1001-125GM E1001-500GM
SPCME1001-125GMEA 180.65 EUR
SPCME1001-125GM SPCME1001-500GM
EDTA dvojsodná sůl dihydrát 99,0 FCC
EDTA dvojsodná sůl dihydrát

Disodium EDTA, Dihydrate, FCC is used as a preservative to inhibit discoloration. Spectrum Chemical manufactured FCC products, including all FCC Disodium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₁₀H₁₄N₂Na₂O₈·2H₂O
Mw: 372.24
Teplota tavenia: 255
Skladovacia teplota: Ambient
Číslo MDL: MFCD00150037
Číslo CAS: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Výsledky špecifikačného testu

Assay (C₁₀H₁₄O₈N₂Na₂2H₂O) 99.0 - 101.0 %
pH of a 1 in 100 Solution 4.3 - 4.7
Calcium (Ca) To Pass Test
Lead (Pb) max. 10 mg/kg
Nitrilotriacetic Acid[N[(HOCOCH₂)₃N] To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR