Tlačiť
Transfer pipettes, Sterilin®
Dodávateľ: THERMO STERILIN
200C 202C
SLIN200CEA 76.51 EUR
SLIN200C SLIN202C
Transfer pipettes, Sterilin®
Pipety Pipety Pasteurovy
These disposable pipettes, manufactured from non toxic LDPE, transfer liquids safely and accurately. Ideal for general laboratory use, sample storage and transport.
  • Integral filling bulb eliminates contamination associated with separate rubber bulbs
  • Excellent transparency and uniform thickness
  • Can be heat sealed for sample storage and transportation
  • Heat-sealed bulbs suitable for storage in the gas phase of liquid nitrogen
  • 1 ml graduated version: Suitable for general laboratory use, storage and transportation of samples
  • 3 ml graduated version: Suitable for
More Product Information
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable