Tlačiť
Stoppers, flange plug caps
Dodávateľ: SIMPORT SCIENTIFIC
T404-3B T404-3N T404-3G T404-3R T404-3Y T404-10N T404-10B T404-10G T404-10R T404-10W T404-10Y
SIMPT404-3BEA 55.68 EUR
SIMPT404-3B SIMPT404-3N SIMPT404-3G SIMPT404-3R SIMPT404-3Y SIMPT404-10N SIMPT404-10B SIMPT404-10G SIMPT404-10R SIMPT404-10W SIMPT404-10Y
Stoppers, flange plug caps
Zátky Zátky PE/LDPE/HDPE
PE Plug type push-in caps and 2-position snap caps are made of polyethylene for test tubes with outside diameters of 12 or 17 mm. Dual position caps offer two possibilities: the closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications.