Tlačiť
Storage canes for Cryovial® tubes
Dodávateľ: SIMPORT SCIENTIFIC
T313
SIMPT313EA 29.6 EUR
SIMPT313
Storage canes for Cryovial® tubes
Slámky kryogenní
These aluminium storage canes can store upto five 1,2 ml or 2 ml tubes in liquid nitrogen containers such as Dewar flasks.