Tlačiť

Dimethylsulfoxid, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

M-Clarity™ quality level = MQ200

D8418-100ML D8418-250ML D8418-50ML
SIALD8418-100MLEA 138.19 EUR
SIALD8418-100ML SIALD8418-250ML SIALD8418-50ML
Dimethylsulfoxid, Sigma-Aldrich®
Dimethylsulfoxid
Vzorec: (CH₃)₂SO
Mw: 78.14
Teplota varu: 189
Teplota tavenia: 18.5
Hustota: 1.101 g/cm³ (20 °C)
Teplota vzplanutia: 87
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002089
Číslo CAS: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR