Tlačiť
PUC18 plasmid DNA
Dodávateľ: Merck
D4154-10UG
SIALD4154-10UGEA 286 EUR
SIALD4154-10UG
PUC18 plasmid DNA
Vektory, plazmidy a knihovny
Plasmids are isolated from host strain E. coli RRI. These plasmids confer ampicillin resistance and complement defects in β-galactosidase in appropriate host strains.
  • Buffered aqueous solution
  • Store at −20 °C
Informace o dodávce: Plasmid DNA is supplied in a solution of 10 mM Tris-HCl, pH 8,0, with 1 mM EDTA.